missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PD1 / PDCD1 / CD279 Protein
A DNA sequence encoding the Homo sapiens (Human) PD1 / PDCD1 / CD279, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7574-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005009.2 | |
PD1 / PDCD1 / CD279 Protein | |
Research Use Only | |
PDCD1 | |
Recombinant Protein | |
E. coli | |
PGWFLDSPDR PWNPPTFSPA LLVVTEGDNA TFTCSFSNTS ESFVLNWYRM SPSNQTDKLA AFPEDRSQPG QDCRFRVTQL PNGRDFHMSV VRARRNDSGT YLCGAISLAP KAQIKESLRA ELRVTERRAE VPTAHPSPSP RPAGQFQTLV | |
Greater than 90% as determined by SDS-PAGE. |
5133 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |