Learn More
enQuireBio™ Recombinant Human NDUFAF2 Protein
A cDNA sequence encoding the NDUFAF2 was constructed and used to recombinantly synthesize the protein.
137.00 € - 7875.00 €
Specifications
Gene ID (Entrez) | 91942 |
---|---|
Name | NDUFAF2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | NDUFAF2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15921639
|
enQuireBio™
QP12814-2UG |
2 μg |
137.00 €
2µg |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15901639
|
enQuireBio™
QP12814-10UG |
10 μg |
212.00 €
10µg |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15911639
|
enQuireBio™
QP12814-1MG |
1 mg |
7875.00 €
1mg |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
91942 | |
Research Use Only | |
NDUFAF2 | |
Human | |
His | |
NDUFAF2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 20% glycerol and 200mM NaCl. |
NDUFAF2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGWSQDLFRA LWRSLSREVK EHVGTDQFGN KYYYIPQYKN WRGQTIREKR IVEAANKKEV DYEAGDIPTE WEAWIRRTRK TPPTMEEILK NEKHREEIKI KSQDFYEKEK LLSKETSEEL LPPPVQTQIK GHASAPYFGK EEPSVAPSST GKTFQPGSWM PRDGKSHNQ | |
Greater than 85.0% as determined by SDS-PAGE. |