missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDUFAF2 Protein

A cDNA sequence encoding the NDUFAF2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12814-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15901639

  • 212.00 € / 10µg
Estimated Shipment: 08-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

91942
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGWSQDLFRA LWRSLSREVK EHVGTDQFGN KYYYIPQYKN WRGQTIREKR IVEAANKKEV DYEAGDIPTE WEAWIRRTRK TPPTMEEILK NEKHREEIKI KSQDFYEKEK LLSKETSEEL LPPPVQTQIK GHASAPYFGK EEPSVAPSST GKTFQPGSWM PRDGKSHNQ
Greater than 85.0% as determined by SDS-PAGE.
NDUFAF2 Protein
Research Use Only
NDUFAF2
Human
His
NDUFAF2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 20% glycerol and 200mM NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions