missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RANGAP1 Monoclonal Antibody (K52049_6F3), Invitrogen™
Mouse Monoclonal Antibody
876.00 €
Specifications
| Antigen | RANGAP1 |
|---|---|
| Clone | K52049_6F3 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: KGAVAMAETL KTLRQVEVIN FGDCLVRSKG AVAIADAIRG GLPKLKELNL SFCEIKRDAA LAVAEAMADK AELEKLDLNG NTLGEEGCEQ LQEVLEGFNM AKVLASLSDD EDEEEEEEGE
RANGAP1 Monoclonal antibody specifically detects RANGAP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| RANGAP1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| C79654; fug1; KIAA1835; MGC20266; mKIAA1835; RAN GTPase activating protein 1; RAN GTPase activating protein 1 a; Ran GTPase activating protein 1 b; Ran GTPase activating protein 1 L homeolog; Ran GTPase activating protein 1 S homeolog; ran GTPase-activating protein 1; RANGAP; RanGAP1; rangap1.L; rangap1.S; rangap1a; rangap1-a; rangap1b; rangap1-b; rna1p; SD; segregation distorter homolog; segregation distortion; XELAEV_18023679mg; XELAEV_18025680mg | |
| Recombinant human RANGAP1 (immunogen range is 251-370). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| RANGAP1 |
| K52049_6F3 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P46060 | |
| 5905 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title