missing translation for 'onlineSavingsMsg'
Learn More

rac Dobutamine Hydrochloride, TRC

Product Code. 17903755
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17903755 10 mg 10mg
30135025 50 mg 50mg
30138097 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17903755 Supplier Toronto Research Chemicals Supplier No. TRCD49444510MG
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49745-95-1
Molecular Formula C18 H23 N O3 . Cl H
Molecular Weight (g/mol) 337.84
Synonym Dobutamine hydrochloride, (RS)-4-[2-[[3-(4-Hydroxyphenyl)-1-methylpropyl]amino]ethyl]benzene-1,2-diol hydrochloride, 1,2-Benzenediol, 4-[2-[[3-(4-hydroxyphenyl)-1-methylpropyl]amino]ethyl]-, hydrochloride (1:1), 1,2-Benzenediol, 4-[2-[[3-(4-hydroxyphenyl)-1-methylpropyl]amino]ethyl]-, hydrochloride (9CI), DL-Dobutamine hydrochloride, Dobutrex, Inotrex, NSC 299583, dl-Dobutamine hydrochloride
IUPAC Name 4-[2-[4-(4-hydroxyphenyl)butan-2-ylamino]ethyl]benzene-1,2-diol;hydrochloride
SMILES Cl.CC(CCc1ccc(O)cc1)NCCc2ccc(O)c(O)c2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H23NO3.ClH/c1-13(2-3-14-4-7-16(20)8-5-14)19-11-10-15-6-9-17(21)18(22)12-15;/h4-9,12-13,19-22H,2-3,10-11H2,1H3;1H
Formula Weight 337.1445
Chemical Name or Material Dobutamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.