missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB31 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
149.35 € - 422.30 €
Specifications
| Antigen | RAB31 |
|---|---|
| Concentration | 0.17 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16815724
|
Proteintech
16182-1-AP-20UL |
20 μL |
149.35 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16805724
|
Proteintech
16182-1-AP-150UL |
150 μL |
422.30 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody can bind RAB31 specifically, and not react RAB22a.
Small GTP-binding proteins of the RAB family, such as RAB31, play essential roles in vesicle and granule targeting.[supplied by OMIM, Jul 2009].Specifications
| RAB31 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| Q13636, Q6GQP4, Q921E2 | |
| 106572, 11031, 246324 | |
| Peptide | |
| Primary | |
| -20°C | |
| RAB31 |
| 0.17 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Rab22B, RAB31, Ras related protein Rab 22B, Ras related protein Rab 31 | |
| Rab31 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title