missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB31 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16182-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody can bind RAB31 specifically, and not react RAB22a.
Small GTP-binding proteins of the RAB family, such as RAB31, play essential roles in vesicle and granule targeting.[supplied by OMIM, Jul 2009].Specifications
| RAB31 | |
| Polyclonal | |
| Unconjugated | |
| RAB31 | |
| Rab22B, RAB31, Ras related protein Rab 22B, Ras related protein Rab 31 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 106572, 11031, 246324 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13636, Q6GQP4, Q921E2 | |
| Rab31 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction