Learn More
(R,R)-Moxifloxacin Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
(R,R)-Moxifloxacin Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 1-cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aR,7aR)-octahydro-6H-pyrrolo [3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic acid, monohydrochloride |
| CAS | 1394029-14-1 |
| Analyte or Component Names | 1-cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aR,7aR)-octahydro-6H-pyrrolo [3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic acid, monohydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H24 F N3 O4 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3-Quinolinecarboxylic acid, 1-cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aR,7aR)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-, hydrochloride (1:1), 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aR,7aR)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic acid monohydrochloride, R,R-Moxifloxacin hydrochloride, (R,R)-Moxifloxacin hydrochloride, Moxifloxacin hydrochloride impurity G |
| InChI Formula | InChI=1S/C21H24FN3O4.ClH/c1-29-20-17-13(19(26)14(21(27)28)9-25(17)12-4-5-12)7-15(22)18(20)24-8-11-3-2-6-23-16(11)10-24;/h7,9,11-12,16,23H,2-6,8,10H2,1H3,(H,27,28);1H/t11-,16+;/m1./s1 |
| SMILES | Cl.COc1c(N2C[C@H]3CCCN[C@H]3C2)c(F)cc4C(=O)C(=CN(C5CC5)c14)C(=O)O |
| IUPAC Name | 7-[(4aR,7aR)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid;hydrochloride |
| Show More |