Learn More
(R)-3-(Carbamoylmethyl)-5-methylhexanoic Acid, Mikromol™
Description
(R)-3-(Carbamoylmethyl)-5-methylhexanoic Acid
Specifications
Specifications
| Chemical Name or Material | (R)-3-(Carbamoylmethyl)-5-methylhexanoic Acid |
| CAS | 181289-33-8 |
| Analyte or Component Names | (R)-3-(Carbamoylmethyl)-5-methylhexanoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H17 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (3R)-3-(2-amino-2-oxoethyl)-5-methylhexanoic acid, Hexanoic acid, 3-(2-amino-2-oxoethyl)-5-methyl-, (R)-, (3R)-3-(2-Amino-2-oxoethyl)-5-methylhexanoic acid, (R)-(-)-3-(Carbamoylmethyl)-5-methylhexanoic acid, (R)-3-(Carbamoylmethyl)-5-methylhexanoic acid |
| InChI Formula | InChI=1S/C9H17NO3/c1-6(2)3-7(4-8(10)11)5-9(12)13/h6-7H,3-5H2,1-2H3,(H2,10,11)(H,12,13)/t7-/m1/s1 |
| SMILES | CC(C)C[C@H](CC(=O)N)CC(=O)O |
| IUPAC Name | (3R)-3-(2-amino-2-oxoethyl)-5-methylhexanoic acid |
| Show More |