Learn More
Quinapyramine Dimethosulfate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Quinapyramine Dimethosulfate |
| CAS | 3270-78-8 |
| Analyte or Component Names | Quinapyramine Dimethosulfate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H21 N6 . C H4 O4 S . C H3 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Quinolinium, 4-amino-6-[(1,2-dihydro-2-imino-1,6-dimethyl-4-pyrimidinyl)amino]-1,2-dimethyl-, methyl sulfate, methyl sulfate (1:1:1), Quinaldinium, 4-amino-6-[(2-amino-1,6-dimethylpyrimidinium-4-yl)amino]-1-methyl-, bis(methyl sulfate) (8CI), Quinolinium, 4-amino-6-[(2-amino-1,6-dimethyl-4(1H)-pyrimidinylidene)amino]-1,2-dimethyl-, methyl sulfate, mono(methyl sulfate) (9CI), Sulfuric acid, monomethyl ester, compd. with 4-amino-6-[(2-amino-1,6-dimethyl-4(1H)-pyrimidinylidene)amino]-1,2-dimethylquinolinium methyl sulfate (1:1) (9CI), Antrycide, Antrycide methyl sulfate, M 7555, Quinapyramine, Quinapyramine methosulfate, Quinapyramine methyl sulfate, Quinapyramine sulfate |
| InChI Formula | InChI=1S/C17H20N6.2CH4O4S/c1-10-7-14(18)13-9-12(5-6-15(13)22(10)3)20-16-8-11(2)23(4)17(19)21-16;2*1-5-6(2,3)4/h5-9,18H,1-4H3,(H2,19,20,21);2*1H3,(H,2,3,4) |
| Solution Type | Neat |
| SMILES | COS(=O)(=O)O.COS(=O)(=O)[O-].CN1C(=C\C(=N\c2ccc3c(c2)c(N)cc(C)[n+]3C)\N=C1N)C |
| Show More |