Learn More
Pyrametostrobin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30325278
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325278 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30325278 Supplier Dr. Ehrenstorfer Supplier No. DREA16599000AL100
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Pyrametostrobin
CAS 915410-70-7
Analyte or Component Names Pyrametostrobin
Shipping Condition +20°C
Molecular Formula C21 H23 N3 O4
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Carbamic acid, N-[2-[[(1,4-dimethyl-3-phenyl-1H-pyrazol-5-yl)oxy]methyl]phenyl]-N-methoxy-, methyl ester, Pyrametostrobin, SYP 4155
InChI Formula InChI=1S/C21H23N3O4/c1-15-19(16-10-6-5-7-11-16)22-23(2)20(15)28-14-17-12-8-9-13-18(17)24(27-4)21(25)26-3/h5-13H,14H2,1-4H3
SMILES CON(C(=O)OC)c1ccccc1COc2c(C)c(nn2C)c3ccccc3
IUPAC Name methyl N-[2-[(2,4-dimethyl-5-phenylpyrazol-3-yl)oxymethyl]phenyl]-N-methoxycarbamate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 381.43
Quantity 1 mL
Formula Weight 381.1689 g/mol