Learn More
Pure Water Density Standard; UKAS ISO/IEC17025 and ISO 17034 certified, nominal density values: 0.9982 g/mL (20°C),0.9970 g/mL (25°C), Triple pack (3 x 60mL), Paragon Scientific™
Click to view available options
Quantity:
2 x 60 mL
Unit Size:
Pack of 3
Description
- Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in accordance with primary level ASTM D1480 methodology
- The reagent water meets the definition of Type II in the Specification ASTM D1193
- Supplied in tamper evident packaging with a 12 month shelf life
- Available as single bottle packs or triple pack kits
- Manufactured in the United Kingdom
Specifications
Specifications
| Quantity | 2 x 60 mL |
Nominal values quoted. Certified Values will vary from batch to batch.