missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSENEN/PEN2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
213.00 € - 462.00 €
Specifications
| Antigen | PSENEN/PEN2 |
|---|---|
| Dilution | Western Blot 1:1000-1:2500, Immunoprecipitation 1:50-1:100 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18243650
|
Novus Biologicals
NB300-227SS |
0.025 mL |
213.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18024784
|
Novus Biologicals
NB300-227 |
0.1 mL |
462.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSENEN/PEN2 Polyclonal specifically detects PSENEN/PEN2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| PSENEN/PEN2 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Growth and Development, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| Whole antisera with 0.05% Sodium Azide | |
| gamma-secretase subunit PEN-2, MSTP064, PEN-2, PEN2hematopoietic stem/progenitor cells protein MDS033, presenilin enhancer 2 homolog (C. elegans), Presenilin enhancer protein 2 | |
| PSENEN | |
| IgG | |
| Unpurified | |
| Human and mouse neuroblastoma (N2a) cells. |
| Western Blot 1:1000-1:2500, Immunoprecipitation 1:50-1:100 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse | |
| Q9NZ42 | |
| 55851 | |
| A synthetic peptide corresponding to the C-terminus of human PEN2 (within residues 50-101). [UniProt# Q9NZ42] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 12 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title