missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSENEN/PEN2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB300-227
This item is not returnable.
View return policy
Description
PSENEN/PEN2 Polyclonal specifically detects PSENEN/PEN2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| PSENEN/PEN2 | |
| Polyclonal | |
| Western Blot 1:1000-1:2500, Immunoprecipitation 1:50-1:100 | |
| Q9NZ42 | |
| PSENEN | |
| A synthetic peptide corresponding to the C-terminus of human PEN2 (within residues 50-101). [UniProt# Q9NZ42] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human and mouse neuroblastoma (N2a) cells. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| gamma-secretase subunit PEN-2, MSTP064, PEN-2, PEN2hematopoietic stem/progenitor cells protein MDS033, presenilin enhancer 2 homolog (C. elegans), Presenilin enhancer protein 2 | |
| Rabbit | |
| 12 kDa | |
| 0.1 mL | |
| Growth and Development, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 55851 | |
| Human, Mouse | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction