missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSCA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
127.00 € - 387.00 €
Specifications
| Antigen | PSCA |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18669694
|
Novus Biologicals
NBP3-21421-100ul |
100 μg |
387.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605194
|
Novus Biologicals
NBP3-21421-25ul |
25 μg |
127.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSCA Polyclonal antibody specifically detects PSCA in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| PSCA | |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, Prostate Cancer, Stem Cell Markers | |
| 0.2um filtered solution in PBS | |
| 8000 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| PRO232, prostate stem cell antigen | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PSCA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title