Learn More
Proxymetacaine N-Oxide Dihydrochloride, Mikromol™
Description
Proxymetacaine N-Oxide Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | Proxymetacaine N-Oxide Dihydrochloride |
| CAS | 2731368-99-1 |
| Analyte or Component Names | Proxymetacaine N-Oxide Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H26 N2 O4 . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Proxymetacaine N-Oxide Dihydrochloride |
| InChI Formula | InChI=1S/C16H26N2O4.2ClH/c1-4-10-21-15-8-7-13(12-14(15)17)16(19)22-11-9-18(20,5-2)6-3;;/h7-8,12H,4-6,9-11,17H2,1-3H3;2*1H |
| SMILES | Cl.Cl.CCCOc1ccc(cc1N)C(=O)OCC[N+]([O-])(CC)CC |
| IUPAC Name | 2-(3-amino-4-propoxybenzoyl)oxy-N,N-diethylethanamine oxide;dihydrochloride |
| Show More |