Learn More
Protionamide Sulfoxide, Mikromol™
Product Code. 30051578
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. 30051578
Brand: Mikromol MM0844.010025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Protionamide Sulfoxide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Protionamide Sulfoxide
CAS 62178-61-4
Analyte or Component Names Protionamide Sulfoxide
Shipping Condition Ice pack (-20°C)
Molecular Formula C9 H12 N2 O S
Certifications/Compliance ISO 9001
Synonym Protionamide Sulfoxide
InChI Formula InChI=1S/C9H12N2OS/c1-2-3-8-6-7(4-5-11-8)9(10)13-12/h4-6H,2-3,10H2,1H3
SMILES CCCc1cc(ccn1)C(=S=O)N
IUPAC Name (2-propylpyridin-4-yl)-sulfinylmethanamine
Molecular Weight (g/mol) 196.27
Quantity 25 mg
Formula Weight 196.067 g/mol