Learn More
Propiomazine Maleate, LoGiCal
Spezifikation
Spezifikation
| Chemical Name or Material | Propiomazine Maleate |
| CAS | 3568-23-8 |
| Analyte or Component Names | Propiomazine Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H24 N2 O S . C4 H4 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-Propanone, 1-[10-[2-(dimethylamino)propyl]-10H-phenothiazin-2-yl]-, (2Z)-2-butenedioate (1:1), 1-Propanone, 1-[10-[2-(dimethylamino)propyl]-10H-phenothiazin-2-yl]-, (Z)-2-butenedioate (1:1), 1-Propanone, 1-[10-[2-(dimethylamino)propyl]phenothiazin-2-yl]-, maleate (1:1) (8CI), Dorevan, Dorevane, Indorm, Phenoctyl, Propavan, Propiomazine maleate, Propiomazine monomaleate |
| InChI Formula | InChI=1S/C20H24N2OS.C4H4O4/c1-5-18(23)15-10-11-20-17(12-15)22(13-14(2)21(3)4)16-8-6-7-9-19(16)24-20;5-3(6)1-2-4(7)8/h6-12,14H,5,13H2,1-4H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| Solution Type | Neat |
| SMILES | CCC(=O)c1ccc2Sc3ccccc3N(CC(C)N(C)C)c2c1.OC(=O)\C=C/C(=O)O |
| Mehr anzeigen |