Learn More
Propamidine Diisetionate, Mikromol™
Description
Propamidine Diisetionate
Specifications
Specifications
| Chemical Name or Material | Propamidine Diisetionate |
| CAS | 140-63-6 |
| Analyte or Component Names | Propamidine Diisetionate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H20 N4 O2 . 2 C2 H6 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ethanesulfonic acid, 2-hydroxy-, compd. with 4,4'-[1,3-propanediylbis(oxy)]bis[benzenecarboximidamide] (2:1), Benzamidine, 4,4'-(trimethylenedioxy)di-, diisethionate (6CI), Ethanesulfonic acid, 2-hydroxy-, compd. with 4,4'-(trimethylenedioxy)dibenzamidine (2:1) (8CI), Ethanesulfonic acid, 2-hydroxy-, compd. with 4,4'-(trimethylenedioxy)dibenzamidine (7CI), Benzamidine, 4,4'-(trimethylenedioxy)di-, bis(2-hydroxyethanesulfonate) (8CI), Benzenecarboximidamide, 4,4'-[1,3-propanediylbis(oxy)]bis-, bis(2-hydroxyethanesulfonate) (9CI), Brolene Drops, M and B 782, Propamidine diisethionate, Propamidine isethionate, p,p'-(Trimethylenedioxy)dibenzamidine isethionate, 4,4'-Diamidino-alpha,omega-diphenoxypropane isethionate, p,p'-(Trimethylenedioxy)dibenzamidine bis(beta-hydroxyethanesulfonate) |
| InChI Formula | InChI=1S/C17H20N4O2.2C2H6O4S/c18-16(19)12-2-6-14(7-3-12)22-10-1-11-23-15-8-4-13(5-9-15)17(20)21;2*3-1-2-7(4,5)6/h2-9H,1,10-11H2,(H3,18,19)(H3,20,21);2*3H,1-2H2,(H,4,5,6) |
| SMILES | NC(=N)c1ccc(OCCCOc2ccc(cc2)C(=N)N)cc1.OCCS(=O)(=O)O.OCCS(=O)(=O)O |
| IUPAC Name | 4-[3-(4-carbamimidoylphenoxy)propoxy]benzenecarboximidamide;2-hydroxyethanesulfonic acid |
| Show More |