Learn More
Prochlorperazine Sulfone N1,N4-Dioxide Dihydrochloride (Prochlorperazine N1,N4,S,S-Tetraoxide Dihydrochloride), Mikromol™
Description
Prochlorperazine Sulfone N1,N4-Dioxide Dihydrochloride (Prochlorperazine N1,N4,S,S-Tetraoxide Dihydrochloride)
Specifications
Specifications
| Chemical Name or Material | Prochlorperazine Sulfone N1,N4-Dioxide Dihydrochloride |
| CAS | 2727065-75-8 |
| Analyte or Component Names | Prochlorperazine Sulfone N1,N4-Dioxide Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H24 Cl N3 O4 S . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Prochlorperazine Sulfone N1,N4-Dioxide Dihydrochloride, Prochlorperazine N1,N4,S,S-Tetraoxide Dihydrochloride |
| InChI Formula | InChI=1S/C20H24ClN3O4S.2ClH/c1-23(25)11-13-24(26,14-12-23)10-4-9-22-17-5-2-3-6-19(17)29(27,28)20-8-7-16(21)15-18(20)22;;/h2-3,5-8,15H,4,9-14H2,1H3;2*1H |
| SMILES | Cl.Cl.C[N+]1([O-])CC[N+]([O-])(CCCN2c3ccccc3S(=O)(=O)c4ccc(Cl)cc24)CC1 |
| IUPAC Name | 2-chloro-10-[3-(4-methyl-1,4-dioxidopiperazine-1,4-diium-1-yl)propyl]phenothiazine 5,5-dioxide;dihydrochloride |
| Show More |