Learn More
Cytiva Processor Plus™ Gel Staining Tray
Product Code. 10350785 Shop All Cytiva Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
10350785 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10350785 Supplier Cytiva Supplier No. 80634391
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Description Processor Plus Gel Staining Tray
Includes Gasket, Splash guard, Magnet plate, Tubing
Material Stainless Steel
Volume (Metric) 125 mL
Dimensions (L x W) 29 x 19 cm