Learn More
Probenecid Isopropyl Ester, Mikromol™
Product Code. 30049934
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
30049934 25 mg 25mg
17963256 100 mg 100mg
30457668 4 x 25 mg 100mg
3 options
Product Code. 30049934
Brand: Mikromol MM0657.030025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Probenecid Isopropyl Ester
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Probenecid Isopropyl Ester
CAS 70190-78-2
Analyte or Component Names Probenecid Isopropyl Ester
Shipping Condition Room Temperature
Molecular Formula C16 H25 N O4 S
Certifications/Compliance ISO 17025
Synonym 4-[(Dipropylamino)sulfonyl]benzoic acid 1-methylethyl ester
InChI Formula InChI=1S/C16H25NO4S/c1-5-11-17(12-6-2)22(19,20)15-9-7-14(8-10-15)16(18)21-13(3)4/h7-10,13H,5-6,11-12H2,1-4H3
SMILES CCCN(CCC)S(=O)(=O)c1ccc(cc1)C(=O)OC(C)C
IUPAC Name propan-2-yl 4-(dipropylsulfamoyl)benzoate
Molecular Weight (g/mol) 327.44
Quantity 25 mg
Formula Weight 327.1504 g/mol