Learn More
Pridinol N-Oxide, Mikromol™
Description
Pridinol N-Oxide
Specifications
Specifications
| Chemical Name or Material | Pridinol N-Oxide |
| CAS | 2724762-69-8 |
| Analyte or Component Names | Pridinol N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H25 N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Pridinol N-Oxide, 1-Piperidinepropanol, α,α-diphenyl-, 1-oxide (ACI), Pridinol N-Oxidee |
| InChI Formula | InChI=1S/C20H25NO2/c22-20(18-10-4-1-5-11-18,19-12-6-2-7-13-19)14-17-21(23)15-8-3-9-16-21/h1-2,4-7,10-13,22H,3,8-9,14-17H2 |
| SMILES | OC(CC[N+]1([O-])CCCCC1)(c2ccccc2)c3ccccc3 |
| IUPAC Name | 3-(1-oxidopiperidin-1-ium-1-yl)-1,1-diphenylpropan-1-ol |
| Show More |