Learn More
Pridinol N-Oxide, Mikromol™
Product Code. 30050979
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050979 25 mg 25mg
30052217 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30050979 Supplier Mikromol Supplier No. MM3493.020025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Pridinol N-Oxide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Pridinol N-Oxide
CAS 2724762-69-8
Analyte or Component Names Pridinol N-Oxide
Shipping Condition Room Temperature
Molecular Formula C20 H25 N O2
Certifications/Compliance ISO 9001
Synonym Pridinol N-Oxide, 1-Piperidinepropanol, α,α-diphenyl-, 1-oxide (ACI), Pridinol N-Oxidee
InChI Formula InChI=1S/C20H25NO2/c22-20(18-10-4-1-5-11-18,19-12-6-2-7-13-19)14-17-21(23)15-8-3-9-16-21/h1-2,4-7,10-13,22H,3,8-9,14-17H2
SMILES OC(CC[N+]1([O-])CCCCC1)(c2ccccc2)c3ccccc3
IUPAC Name 3-(1-oxidopiperidin-1-ium-1-yl)-1,1-diphenylpropan-1-ol
Molecular Weight (g/mol) 311.42
Quantity 25 mg
Formula Weight 311.1885 g/mol