Learn More
Pregabalin EP Impurity D, Mikromol™
Product Code. 30461040
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461040 4 x 25 mg -
30051568 25 mg 25mg
17912876 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30461040 Supplier Mikromol Supplier No. MM1376.0500254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Pregabalin EP Impurity D, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Isopropyl Mandelate
CAS 4118-51-8
Analyte or Component Names Isopropyl Mandelate
Shipping Condition Room Temperature
Molecular Formula C11 H14 O3
Certifications/Compliance ISO 9001
Synonym Propan-2-yl (2RS)-2-Hydroxy-2-phenylacetate (Isopropyl Mandelate), Pregabalin impurity D, Pregabalin Imp. D (EP), alpha-Hydroxy-benzeneacetic acid 1-Methylethyl ester, Propan-2-yl (2RS)-2-Hydroxy-2-phenylacetate, Mandelic acid, isopropyl ester (7CI,8CI), (+/-)-Isopropyl mandelate, (+/-)-Mandelic acid isopropyl ester, Isopropyl 2-hydroxy-2-phenylacetate, Isopropyl mandelate, NSC 6582, Pregabalin Impurity D
InChI Formula InChI=1S/C11H14O3/c1-8(2)14-11(13)10(12)9-6-4-3-5-7-9/h3-8,10,12H,1-2H3
Solution Type Neat
SMILES CC(C)OC(=O)C(O)c1ccccc1
IUPAC Name propan-2-yl 2-hydroxy-2-phenylacetate
Molecular Weight (g/mol) 194.227
Quantity 4 x 25 mg
Formula Weight 194.094 g/mol