Learn More
Pregabalin EP Impurity D, Mikromol™
Description
Pregabalin EP Impurity D
Specifications
Specifications
| Chemical Name or Material | Isopropyl Mandelate |
| CAS | 4118-51-8 |
| Analyte or Component Names | Isopropyl Mandelate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H14 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Propan-2-yl (2RS)-2-Hydroxy-2-phenylacetate (Isopropyl Mandelate), Pregabalin impurity D, Pregabalin Imp. D (EP), alpha-Hydroxy-benzeneacetic acid 1-Methylethyl ester, Propan-2-yl (2RS)-2-Hydroxy-2-phenylacetate, Mandelic acid, isopropyl ester (7CI,8CI), (+/-)-Isopropyl mandelate, (+/-)-Mandelic acid isopropyl ester, Isopropyl 2-hydroxy-2-phenylacetate, Isopropyl mandelate, NSC 6582, Pregabalin Impurity D |
| InChI Formula | InChI=1S/C11H14O3/c1-8(2)14-11(13)10(12)9-6-4-3-5-7-9/h3-8,10,12H,1-2H3 |
| SMILES | CC(C)OC(=O)C(O)c1ccccc1 |
| IUPAC Name | propan-2-yl 2-hydroxy-2-phenylacetate |
| Show More |