missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Praziquantel D11, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Praziquantel D11 is the deuterium labeled Praziquantel, which is an anthelmintic.
Specifications
Specifications
| Chemical Name or Material | Praziquantel D11 |
| CAS | 1246343-36-1 |
| Purity Grade Notes | Research |
| Molecular Formula | C19H13D11N2O2 |
| SMILES | O=C1N2CCC3=C(C=CC=C3)C2CN(C(C4([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C4([2H])[2H])=O)C1 |
| Industry Type | COVID-19-immunoregulation |
| Molecular Weight (g/mol) | 323.47 |
| Quantity | 10 mg |
| Formula Weight | 323.47 |
| Grade | Research |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction