Learn More
Prasedymium solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
Product Code. p-7083811 Shop All Fisher Chemical Products
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Bottle
Unit Size:
100mL
50mL
Product Code. 12907403
Brand: Fisher Chemical S 643 050
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Chemical Identifiers
Molecular Formula Pr
Specifications
Packaging Bottle
Concentration 10 mg/L
Solvent or Matrix Nitric Acid
Quantity 50 mL
Chemical Name or Material Prasedymium