Learn More
Kartell™ Non-Electric Magnetic Stirrer
Click to view available options
Type:
Magnetic Stirrer
Unit Size:
Each
Description
- Operated by water pressure between 15 to 40 psi) or by air pressure
- No electric supply is needed thus eliminating heat, shock and explosion hazard
- Corrosion proof allplastic construction
- Can be used on the bench or clamped to a stand with 12mm diameter
- Outlet tube 3.4mm
- Inlet tube 13.20mm
Specifications
Specifications
| Top Plate Material | ABS |
| Diameter (Metric) | 128.5 mm |
| Height (Metric) | 61.3 mm |
| Speed Control | Water or air pressure |
| Type | Magnetic Stirrer |