Learn More
Potassium Ferrocyanide Solution According to European Pharmacopoeia (EP), Reagecon
BIOPHARMA
Product Code. 11404729 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404729 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11404729 Supplier Reagecon Supplier No. 1069801
  • ONLINE EXCLUSIVE PRICE
    69.95 €
    60.72 € / 100mL
    Save 9.23 €
    13% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity stable product developed and tested for European Pharmacopoeia (EP) applications.
  • European Pharmacopoeia Reagent, Solution
  • High-purity stable product developed and tested for European Pharmacopoeia (EP) applications.
  • These include use in the identification reactions of ions and functional groups (2.3.1) and as a reagent in the performance of tests outlined in several EP monographs.
  • NIST traceable.
  • This Reagecon EP reagent is accurate, stable and reproducible.
  • Stability is proven (either opened or unopened) for the shelf life of this product.
Specifications
Packaging HDPE Bottle
Physical Form Liquid
Technique EP Testing
Quantity 100 mL