missing translation for 'onlineSavingsMsg'
Learn More

Potassium antimonate trihydrate, 94+%

Product Code. p-4407714
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11318328

Brand: Thermo Scientific Alfa Aesar 039524.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12208-13-8 | KSbO3·3H2O

Potassium antimonyl trihydrate is used as a detecting agent for sodium. It is used as a mordant/ fixing agent in leather and textile dying industries. It is also used as a analytical reagent and flux additive for electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12208-13-8
Molecular Formula KSbO3·3H2O
MDL Number MFCD00150439

Specifications

Color White
Physical Form Crystals or Powder
Quantity 50 g
Assay Percent Range >94%
UN Number 1549
Synonym Potassium Pyroantimonate, Potassium Hexahydroxyantimonate
Solubility Information Soluble in water and glycerine. Insoluble in ethanol.
Formula Weight 262.90 (208.85 Anhydrous)
Appearance White powder
Chemical Name or Material Potassium Antimonate Trihydrate
compliance-icons compliance-icons
Product Identifier
  • Potassium antimonate trihydrate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P330-Rinse mouth.
  • P391-Collect spillage.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.