CAS RN 12208-13-8
CAS RN 12208-13-8
IUPAC Name:
antimony(3+) potassium hexahydroxide
Synonyms:
antimony(3+) potassium hexahydroxide
Molecular Weight (g/mol):
262.9
Molecular Formula:
H6KO6Sb
InChi Key:
IDVSLFDKDQVCAP-UHFFFAOYSA-H
SMILES:
[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[K+].[Sb+3]
1
–
2
of
2
results
1
Potassium antimonate trihydrate, 94+%
CAS: 12208-13-8 Molecular Formula: KSbO3·3H2O MDL Number: MFCD00150439
2
Potassium hexahydroxoantimonate(V), Honeywell Fluka™
CAS: 12208-13-8 MDL Number: MFCD00011415