missing translation for 'onlineSavingsMsg'
Learn More

JG Finneran Associates™ Poly Crimp™ Seals

Brand:  JG Finneran Associates™ 5581-11

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10696041

  • 525.00 € / Pack of 1000
Estimated Shipment: 01-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

PTFE/Silicone with Starburst
PTFE, Silicone
11 mm
Clear
1000/Pk.
12 x 32 mm Sample Vials with Crimp Style Finish
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions