Learn More
JG Finneran Associates™ Poly Crimp™ Seals
183.00 € - 535.00 €
Specifications
Color | Clear |
---|---|
Quantity | 1000/Pk. |
Closure Size | 11 mm |
For Use With (Equipment) | 12 x 32 mm Sample Vials with Crimp Style Finish |
Product Code | Brand | Material (Liner) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material (Liner) | Price | Quantity & Availability | |||||
10626991
|
JG Finneran Associates™
5510-11 |
PTFE |
183.00 €
Pack of 1000 |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
10025570
|
JG Finneran Associates™
5570-11 |
PTFE/Silicone with Slit |
535.00 €
Pack of 1000 |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
10696041
|
JG Finneran Associates™
5581-11 |
PTFE/Silicone with Starburst |
525.00 €
Pack of 1000 |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
10391241
|
JG Finneran Associates™
5560-11 |
PTFE/Silicone/PTFE |
327.00 €
Pack of 1000 |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
Specifications
Clear | |
1000/Pk. | |
11 mm | |
12 x 32 mm Sample Vials with Crimp Style Finish |