Learn More
Memmert™ Plug-In Tube Extension for IN110, Straight
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IN110 Incubator
Unit Size:
Each
Description
- Accessory
- Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
- Straight
Specifications
Specifications
| For Use With (Equipment) | IN110 Incubator |
| Type | Plug-in Tube Extension |
| Diameter (Metric) | O.D. 60.3 mm, I.D. 57 mm |