missing translation for 'onlineSavingsMsg'
Learn More

Platinum solution for ICP/MS 10mg/L HCl matrix, Fisher chemical™

Product Code. 12927403 Shop All Fisher Chemical Products
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Bottle
Unit Size:
100mL
50mL
This item is not returnable. View return policy

Product Code. 12927403

Brand: Fisher Chemical S 644 050

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 6-6-7440
Molecular Formula Pt

Specifications

Packaging Bottle
Concentration 10 mg/L
Solvent or Matrix Hydrogen Chloride
Quantity 50 mL
Chemical Name or Material Platinum
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.