missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piroxicam, MedChemExpress
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
Description
Piroxicam (CP-16171) is a non-steroidal anti-inflammatory drugs, acts as a COX inhibitor, with IC50s of 47, 25 μM for human monocyte COX-1 and COX-2, respectively.
Specifications
Specifications
| Chemical Name or Material | Piroxicam |
| CAS | 36322-90-4 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C15H13N3O4S |
| Quantity | 5 g |
| Synonym | CP-16171 |
| Solubility Information | DMSO : 50 mg/mL (150.90 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C(C1=C(O)C2=CC=CC=C2S(N1C)(=O)=O)NC3=NC=CC=C3 |
| Molecular Weight (g/mol) | 331.35 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction