Learn More
Melford PIPES Sodium Salt
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
500g
5kg
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
pKa (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5
Specifications
Specifications
| Chemical Name or Material | PIPES, Sodium Salt |
| CAS | 10010-67-0 |
| Quantity | 500 g |
| Molecular Formula | C8H17N2NaO6S2 |