Learn More
Pipemidic Acid Isopropyl Ester, Mikromol™
Description
Pipemidic Acid Isopropyl Ester
Specifications
Specifications
| Chemical Name or Material | Pipemidic Acid Isopropyl Ester |
| CAS | 2724726-52-5 |
| Analyte or Component Names | Pipemidic Acid Isopropyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H23 N5 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Pipemidic Acid Isopropyl Ester |
| InChI Formula | InChI=1S/C17H23N5O3/c1-4-21-10-13(16(24)25-11(2)3)14(23)12-9-19-17(20-15(12)21)22-7-5-18-6-8-22/h9-11,18H,4-8H2,1-3H3 |
| SMILES | CCN1C=C(C(=O)OC(C)C)C(=O)c2cnc(nc12)N3CCNCC3 |
| IUPAC Name | propan-2-yl 8-ethyl-5-oxo-2-piperazin-1-ylpyrido[2,3-d]pyrimidine-6-carboxylate |
| Show More |