missing translation for 'onlineSavingsMsg'
Learn More

PIK-93, MedChemExpress

Product Code. 16451197
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16451197

Brand: Medchem Express HY12046

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PIK-93 is the first potent, synthetic PI4K (PI4KIIIβ) inhibitor with IC50 of 19 nM, and also inhibits PI3Kγ and PI3Kα with IC50 of 16 nM and 39 nM, respectively.

PIK-93 is the first potent, synthetic PI4K (PI4KIIIβ) inhibitor with IC50 of 19 nM, and also inhibits PI3Kγ and PI3Kα with IC50 of 16 nM and 39 nM, respectively.

Specifications

Chemical Name or Material PIK-93
CAS 593960-11-3
Color White
Purity Grade Notes Research
Molecular Formula C14H16ClN3O4S2
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (256.49 mM; Need ultrasonic)
SMILES CC(NC1=NC(C)=C(S1)C2=CC=C(C(S(=O)(NCCO)=O)=C2)Cl)=O
Molecular Weight (g/mol) 389.88
Formula Weight 389.88
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.