missing translation for 'onlineSavingsMsg'
Learn More

PIAS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen PIAS3
Concentration 0.23 mg/mL
Applications Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16841724
View Documents
Proteintech
13486-1-AP-20UL
20 μL
145.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16831724
View Documents
Proteintech
13486-1-AP-150UL
150 μL
410.00 €
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The PIAS3 antibody detects PIAS3 (68 kDa) as well as its sumoylated form (85 kDa) in some normal mouse tissues, breast and brain tumor cell lines.

The PIAS (protein inhibitor of activated STAT) proteins play a crucial role as transcriptional coregulators in various cellular pathways, including the STAT, p53 and the steroid hormone signaling pathway. The PIAS protein family includes at least five evolutionarily conserved genes, including PIAS3. The major function of the PIAS proteins is the control of gene transcription and can also act as small ubiquitin-like-modifier (SUMO) E3 ligases. PIAS3 binds specifically to STAT3 following the stimulation of STAT3. Increased expression of PIAS3 has been observed in several human cancers, including lung, breast, and brain tumors, but not in anaplastic lymphoma kinase-positive T/null-cell lymphomas, indicating that PIAS3 plays multiple roles in different tissue and cell types.
Specifications

Specifications

PIAS3
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Mouse, Human, Rat
O54714, O70260, Q9Y6X2
10401, 229615, 83614
PIAS3 Fusion Protein Ag4304
Primary
-20°C
PIAS3
0.23 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
E3 SUMO protein ligase PIAS3, PIAS3, ZMIZ5
PIAS3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.