Learn More
Phenyramidol Hydrochloride, Mikromol™
Description
Phenyramidol Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Phenyramidol Hydrochloride |
| CAS | 326-43-2 |
| Analyte or Component Names | Phenyramidol Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H14 N2 O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Abbolexin, Anabloc, Analexin, Betapirin, Elan, Fenyramidol hydrochloride, Firmalgil, IN 511, MJ 505, Miodar, NSC 17777, Phenylramidol hydrochloride, Phenyramidol hydrochloride, Vazalmin, Verbanol, Benzenemethanol, alpha-[(2-pyridinylamino)methyl]-, hydrochloride (1:1), Benzenemethanol, alpha-[(2-pyridinylamino)methyl]-, monohydrochloride (9CI), Benzyl alcohol, alpha-[(2-pyridylamino)methyl]-, hydrochloride (7CI), Benzyl alcohol, alpha-[(2-pyridylamino)methyl]-, monohydrochloride (6CI,8CI), alpha-[(2-Pyridylamino)methyl]benzyl alcohol hydrochloride |
| InChI Formula | InChI=1S/C13H14N2O.ClH/c16-12(11-6-2-1-3-7-11)10-15-13-8-4-5-9-14-13;/h1-9,12,16H,10H2,(H,14,15);1H |
| SMILES | Cl.OC(CNc1ccccn1)c2ccccc2 |
| IUPAC Name | 1-phenyl-2-(pyridin-2-ylamino)ethanol;hydrochloride |
| Show More |