Learn More
Phenylacetylasparagine, Mikromol™
Description
Phenylacetylasparagine
Specifications
Specifications
| Chemical Name or Material | Phenylacetylasparagine |
| CAS | 2752-49-0 |
| Analyte or Component Names | Phenylacetylasparagine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H14 N2 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Phenylacetylasparagine |
| InChI Formula | InChI=1S/C12H14N2O4/c13-10(15)7-9(12(17)18)14-11(16)6-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H2,13,15)(H,14,16)(H,17,18)/t9-/m0/s1 |
| SMILES | NC(=O)C[C@H](NC(=O)Cc1ccccc1)C(=O)O |
| IUPAC Name | (2S)-4-amino-4-oxo-2-[(2-phenylacetyl)amino]butanoic acid |
| Show More |