Learn More
Phentolamine Hydrochloride, Mikromol™
Description
Phentolamine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Phentolamine Hydrochloride |
| CAS | 73-05-2 |
| Analyte or Component Names | Phentolamine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H19 N3 O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Phenol, 3-[[(4,5-dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]-, hydrochloride (1:1), Phenol, 3-[[(4,5-dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]-, monohydrochloride (9CI), Phenol, m-[N-(2-imidazolin-2-ylmethyl)-p-toluidino]-, monohydrochloride (8CI), DNP, Phentolamine chloride, Phentolamine hydrochloride, Regitine hydrochloride, m-[N-(2-Imidazolin-2-ylmethyl)-p-toluidino]phenol hydrochloride |
| InChI Formula | InChI=1S/C17H19N3O.ClH/c1-13-5-7-14(8-6-13)20(12-17-18-9-10-19-17)15-3-2-4-16(21)11-15;/h2-8,11,21H,9-10,12H2,1H3,(H,18,19);1H |
| SMILES | Cl.Cc1ccc(cc1)N(CC2=NCCN2)c3cccc(O)c3 |
| IUPAC Name | 3-[N-(4,5-dihydro-1H-imidazol-2-ylmethyl)-4-methylanilino]phenol;hydrochloride |
| Show More |