Learn More
Eleven Components Phenol Standard solution, 2000μg/mL of each, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Eleven Components Standard Solution |
| Packaging | Ampoule |
| Analyte or Component Names | 4-Chloro-3-methylphenol (CAS:59-50-7), 2-Chlorophenol (CAS:95-57-8), 2,4- Dichlorophenol (CAS:120-83-2), 2,4-Dimethylphenol (CAS:105-67-9), 2,4-Dinitrophenol (CAS:51-28 -5), 2-Methyl-4,6-dinitrophenol (CAS:534-52-1), 2-Nitrophenol (CAS:88-75-5), 4-Nitrophenol (CAS:100-02-7), Pentachlorophenol (CAS:87-86-5), Phenol (CAS:108-95-2), 2,4,6-Trichlorophenol (CAS:88-06-2) |
| Concentration | 2000 μg/mL |
| UN Number | 1593 |
| Type | Organic Multi-component Standard Solution |
| Element | 4-Chloro-3-methylphenol, 2-Chlorophenol, 2,4- Dichlorophenol, 2,4-Dimethylphenol, 2,4-Dinitrophenol, 2-Methyl-4,6-dinitrophenol, 2-Nitrophenol, 4-Nitrophenol, Pentachlorophenol, Phenol, 2,4,6-Trichlorophenol |
| Solvent or Matrix | Dichloromethane |
| Quantity | 1 mL |
Safety and Handling
Product Identifier
- Standard solution 11 Components in dichloromethane
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H351-Suspected of causing cancer.
- H412-Harmful to aquatic life with long lasting effects.
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .