missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH Buffer Solution pH 6.865, Mettler Toledo™
Shop All Mettler Toledo Products
Click to view available options
Quantity:
6 x 250 mL
Unit Size:
1500mL
Specifications
Specifications
| pH | 6.865 |
| Physical Form | Liquid |
| Quantity | 6 x 250 mL |
| Traceability to NIST | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction