missing translation for 'onlineSavingsMsg'
Learn More

pH Buffer Solution pH 6.00, Carl Roth™

Product Code. 10152013
Encompass
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10152013 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10152013 Supplier Carl Roth Supplier No. T184.2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Absorbance 500mL
Color Colorless
pH 6.00
Physical Form Liquid
Quantity 500 mL
Formulation Phosphate
Odor Characteristic
Packaging Plastic
Traceability to NIST Yes
Solubility Information Fully miscible in water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.