missing translation for 'onlineSavingsMsg'
Learn More

pH Buffer Solution, pH 12, Carl Roth™

Product Code. 10019892
Encompass
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
This item is not returnable. View return policy

Product Code. 10019892

Brand: Carl Roth T189.1

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Keeps pH at a nearly constant value. Carl Roth pH Buffer Solution, pH 12 can be used in a wide variety of chemical applications

Buffer solution: pH 12.00 ±0.05 (20 °C)

Ready to use

Glycine/Caustic soda lye/Sodium chloride

Specifications

Melting Point 0°C
Color Colorless
pH 12
Physical Form Liquid
Quantity 1 L
Solution Type pH Buffer
Vapor Pressure 23hPa
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.