Learn More
PFI-1, MedChemExpress
Product Code. 16497126
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16497126
Brand: Medchem Express HY16586
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress PFI-1 is a selective BET (bromodomain-containing protein) inhibitor for BRD4 with IC50 of 0.22 μM in a cell-free assay.
PFI-1 is a selective BET (bromodomain-containing protein) inhibitor for BRD4 with IC50 of 0.22 μM in a cell-free assay.
Specifications
Chemical Name or Material PFI-1
CAS 1403764-72-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H17N3O4S
Quantity 100 mg
Solubility Information DMSO : 33.33 mg/mL (95.94 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC=C1OC)(NC2=CC3=C(NC(N(C)C3)=O)C=C2)=O
Molecular Weight (g/mol) 347.39
Formula Weight 347.39
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease