Learn More
Perphenazine Sulfoxide N1-Oxide (Perphenazine N1,S-Dioxide), Mikromol™
Description
Perphenazine Sulfoxide N1-Oxide (Perphenazine N1,S-Dioxide)
Specifications
Specifications
| Chemical Name or Material | Perphenazine Sulfoxide N1-Oxide |
| CAS | 2731414-17-6 |
| Analyte or Component Names | Perphenazine Sulfoxide N1-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H26 Cl N3 O3 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Perphenazine Sulfoxide N1-Oxide |
| InChI Formula | InChI=1S/C21H26ClN3O3S/c22-17-6-7-21-19(16-17)24(18-4-1-2-5-20(18)29(21)28)9-3-8-23-10-12-25(27,13-11-23)14-15-26/h1-2,4-7,16,26H,3,8-15H2 |
| SMILES | OCC[N+]1([O-])CCN(CCCN2c3ccccc3S(=O)c4ccc(Cl)cc24)CC1 |
| IUPAC Name | 2-[4-[3-(2-chloro-5-oxophenothiazin-10-yl)propyl]-1-oxidopiperazin-1-ium-1-yl]ethanol |
| Show More |